Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vitamin K1, MP Biomedicals™

Catalog No. ICN10328305 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10328305 5 g
ICN10328301 1 g
ICN10328310 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10328305 Supplier MP Biomedicals, Inc Supplier No. 0210328305
Add to Cart
Add to Cart

Chemical Identifiers

CAS 84-80-0
Molecular Formula C31H46O2
Molecular Weight (g/mol) 450.707
InChI Key MBWXNTAXLNYFJB-NKFFZRIASA-N
Synonym vitamin k1, phytonadione, phylloquinone, phytylmenadione, phyllochinon, phytomenadione, alpha-phylloquinone, 3-phytylmenadione, konakion, 2-methyl-3-phytyl-1,4-naphthoquinone
PubChem CID 5284607
ChEBI CHEBI:18067
IUPAC Name 2-methyl-3-[(E,7R,11R)-3,7,11,15-tetramethylhexadec-2-enyl]naphthalene-1,4-dione
SMILES CC1=C(C(=O)C2=CC=CC=C2C1=O)CC=C(C)CCCC(C)CCCC(C)CCCC(C)C

Specifications

Color Yellow
Density 0.984
Quantity 5 g
Formula Weight 450.68
Physical Form Viscous Liquid
Chemical Name or Material Vitamin K1
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.