Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vitrin Polyclonal Antibody
GREENER_CHOICE

Vitrin Polyclonal Antibody

Supplier:  Thermo Scientific PA524009

Catalog No. PIPA524009


Only null left
Add to Cart

Description

Description

Vitrin Polyclonal Antibody for Western Blot, IHC (P), Flow

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Vitrin
Polyclonal
Unconjugated
VIT
UNQ647/PRO1277; VIT; Vitrin
Rabbit
Antigen affinity chromatography, Protein A
RUO
5212
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.4
Q6UXI7
VIT
KLH conjugated synthetic peptide between 567-595 amino acids from the C-terminal region of human VITRN.
400 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.