Learn More
Description
- The VitroEase™ Cryo-EM Training Kit has been designed to provide the necessary components to get started with cryo-EM analysis of protein samples
- Sample preparation is a major bottleneck in cryo-electron microscopy (cryo-EM) single particle analysis (SPA)
- Currently, sample optimization is often performed in a non-systematic way, extending optimization time over multiple days or even weeks
- Kit includes a positive control protein, apoferritin, that has been designed specifically to meet the stringent needs for high protein purity, minimal protein aggregation, and intact protein structure
- Commonly used accessories for grid preparation are also included such as tweezers for manipulation, grids, and grid boxes for storage
- Detailed step-by-step instructions guide the user through the lengthy vitrification process
- Together the control and components may be used as a comparison for success for new users or for quality control for expert users
- Grids and accessories are provided by trusted sources
Specifications
Specifications
| Content And Storage | Sufficient for 25 grid preparations. Contains VitroEase Apoferritin Standard (Cat. No. A51362), tweezers, grids, and grid boxes. |
| For Use With (Application) | Cryo-EM SPA |
| For Use With (Equipment) | Cryo-Electron Microscopes |
| Quantity | 25 Preparations |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.