Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vitronectin Antibody (019), Alexa Fluor™ 594, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290648AF594
Description
Vitronectin Monoclonal specifically detects Vitronectin in Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Vitronectin | |
Monoclonal | |
Alexa Fluor 594 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
19 | |
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
complement S-protein, epibolin, serum spreading factor, Serum-spreading factor, somatomedin B, S-protein, V75, vitronectin, vitronectin (serum spreading factor, somatomedin B, complement S-protein), VN, VNT | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Vitronectin (NP_035837.1; Met1-Lys478). | |
0.1 mL | |
Immunology, Innate Immunity | |
7448 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction