Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vitronectin Recombinant Rabbit Monoclonal Antibody (2V2V8)
Vitronectin Recombinant Rabbit Monoclonal Antibody (ARC0313)
Supplier: Thermo Scientific MA535597
Description
Vitronectin Recombinant Monoclonal Antibody for Western Blot

Specifications
Vitronectin | |
Recombinant Monoclonal | |
0.5 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P04004 | |
VTN | |
A synthetic peptide corresponding to a sequence within amino acids 379-478 of human Vitronectin (P04004). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA, Western Blot | |
2V2V8 | |
Unconjugated | |
VTN | |
Aa1018; AI256434; complement S-protein; epibolin; extracellular matrix protein; glycoprotein 66; serum spreading factor; serum-spreading factor; somatomedin B; Somatomedin-B; S-protein; V75; Vitronectin; vitronectin (serum spreading factor, somatomedin B, complement S-protein); Vitronectin V10 subunit; Vitronectin V65 subunit; VN; VNT; VTN | |
Rabbit | |
Affinity Chromatography | |
RUO | |
7448 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction