Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vitronectin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | Vitronectin |
---|---|
Concentration | 0.33 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-0515
![]() ![]() |
Proteintech Group Inc
158331AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Vitronectin is a plasma glycoprotein that circulates in the blood. Vitronectin is circulating as a mixture of both 75 kDa and 65 kDa forms. Vitronectin is a major cell adhesive glycoprotein and is a common component of extracellular matrix and plasma. It competes effectively with other plasma proteins and is often involved in cell attachment, regulation of blood coagulation and immune responses. It has similar tissue distribution to fibronectin and also its integrin receptor recognises fibronectin.Specifications
Vitronectin | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
P04004, P29788 | |
22370, 29169, 7448 | |
Vitronectin Fusion Protein Ag8443 | |
Primary | |
-20°C | |
VTN |
0.33 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
S protein, Serum spreading factor, V75, vitronectin, VN, VNT, VTN | |
Vtn | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title