Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vitronectin Recombinant Rabbit Monoclonal Antibody (019)
GREENER_CHOICE

Catalog No. PIMA529842
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA529842 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA529842 Supplier Thermo Scientific Supplier No. MA529842
Add to Cart
Add to Cart

Vitronectin Recombinant Rabbit Monoclonal Antibody (19)

Vitronectin Recombinant Monoclonal Antibody for IHC (P), ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen Vitronectin
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Recombinant Monoclonal
Clone 019
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene VTN
Gene Accession No. P29788
Gene Alias Aa1018; AI256434; complement S-protein; epibolin; extracellular matrix protein; glycoprotein 66; serum spreading factor; serum-spreading factor; somatomedin B; Somatomedin-B; S-protein; V75; Vitronectin; vitronectin (serum spreading factor, somatomedin B, complement S-protein); Vitronectin V10 subunit; Vitronectin V65 subunit; VN; VNT; VTN
Gene Symbols VTN
Host Species Rabbit
Immunogen Recombinant Mouse Vitronectin/VTN protein (Met1-Lys478).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22370
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.