Learn More
QuidelOrtho VITROS™ Immunodiagnostic Products B·R·A·H·M·S PCT Reagent
Catalog No. OCD6197735
Change view
Click to view available options
Quantity:
100 Tests
Catalog No. Quantity
OCD6197735 100 Tests
1 options
Catalog No. OCD6197735 Supplier QuidelOrtho Supplier No. 6197735
 Restricted Item
Add to Cart
Add to Cart
QuidelOrtho B·R·A·H·M·S PCT is a biomarker for early bacterial infection diagnosis and antibiotic stewardship
Test delivers:
  • High sensitivity and specificity for bacterial infection enables therapeutic decision making
  • Results that are ready to be delivered to a clinicians with 96.5% First Pass Yield (without user intervention)
  • Analytical performance: LOD at 0.007 ng/mL, LOQ (20% CV, observed) at 0.013 ng/mL
  • Fast turnaround time: 24 minutes to first result
  • Reliable solution: Not impacted by biotin interference
Indicated as an aid to be used in conjunction with clinical evaluation for:
  • Risk assessment of critically ill patients on their first day of ICU admission for progression to severe sepsis and septic shock
  • Assessing the cumulative 28-day risk of all-cause mortality for patients diagnosed with severe sepsis or septic shock in the ICU or when obtained in the emergency department or other medical wards prior to ICU admission, using a change in PCT level over time
  • Decision making on antibiotic therapy for patients with suspected or confirmed lower respiratory tract infections (LRTI) defined as community-acquired pneumonia (CAP), acute bronchitis, and acute exacerbation of chronic obstructive pulmonary disease (AECOPD) – in an inpatient setting or an emergency department
  • Decision making on antibiotic discontinuation for patients with suspected or confirmed sepsis
Indicated for use with the VITROS™ ECi/ECiQ/3600 Immunodiagnostic Systems and the VITROS™ 5600/XT 7600 Integrated Systems.
Specifications
Type Reagent
Quantity 100 Tests
Range 0.030 to 100 ng/mL (0.030 to 100 μg/L)
Sample Size 30 μL
Test Time 24 min.