Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BRANDTECH™ VITLAB™ VITsafe™ Wide-Mouth Wash Bottle
Shop All BrandTech Scientific Products
Click to view available options
Chemical Label:
Acetone
Distilled Water
Ethanol
Heptane
Xylene
i-Hexane
Description
- Chemical name in German, English, French and Spanish
- Chemical formula
- CAS Number, hazard pictogram, signal word
- Risk Phrases (H phrases), safety phrases (P phrases)
- NFPA Code
- Fine tip polypropylene tube provides directional accuracy
- Colored cap allows easier identification

Specifications
Specifications
Capacity (Metric) | 500 mL |
Capacity (English) | 16.9 oz. |
Color | Red |
For Use With (Application) | Heptane |
Material | LDPE, Polypropylene |
Length (Metric) | 166 mm |
Packaging | Pack |
Quantity | 6 |
Sterility | Non-sterile |
Width (Metric) | 76 mm |
Show More |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction