Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Stedim Biotech™ Vivacon 500 125kDa

Catalog No. 02226206 Shop All Sartorius Products
Change view
Click to view available options
Quantity:
100 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-226-206 100 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-226-206 Supplier Sartorius Stedim Biotech™ Supplier No. VN01H82
Add to Cart
Add to Cart

Vivacon 500 125kDa are centrifugal units for fast DNA concentration and buffer exchange of 500 μL samples.

  • The product of choice for protein fractionation applications.
  • The use of Hydrosart™ membranes results in high recoveries from dilute samples.
  • Complete and highly reproducible recovery of the concentrate.
  • Reverse spinning step capacity.
  • Concentration of up to 0.5 mL genomic DNA or peptide samples.
  • Low binding material.
  • MWCO printed on device.
  • Selected cut offs available sterile and DNA-free for things like forensic applications.
  • qPCR assay over 55 cycles with a verified limit of detection <2 pg
TRUSTED_SUSTAINABILITY

Specifications

Color Transparent
Volume (Metric) 0.5 mL
Quantity 100 Pk.
Product Line Centrifugal ultrafiltration device
Sterility Sterile
Material Polypropylene
Closure Type Flat Cap
Autoclavable Not Autoclavable
Closure Included Yes
Disposable Single-use
Product Type Centrifugal Units
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.