Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Vivacon™ 500 DNA Concentrator

Catalog No. 14558351 Shop All Sartorius Products
Change view
Click to view available options
MWCO:
10 kDa
100 kDa
2 kDa
30 kDa
50 kDa
Quantity:
100 Pack
25 Pack
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. MWCO Quantity
14-558-351 50 kDa 100 Pack
14-558-350 50 kDa 25 Pack
14-558-343 10 kDa 25 Pack
14-558-344 10 kDa 100 Pack
14-558-345 30 kDa 25 Pack
14-558-353 100 kDa 25 Pack
14-558-347 30 kDa 100 Pack
14-558-355 100 kDa 100 Pack
14-558-359 2 kDa 100 Pack
14-558-358 2 kDa 25 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 14-558-351 Supplier Sartorius Supplier No. VN01H32
Add to Cart
Add to Cart

For up to 500 μL, Vivacon™ 500 are centrifugal units for fast DNA concentration and buffer exchange of 500 μL samples. They are also the product of choice for protein fractionation applications. The use of Hydrosart™ membranes results in high recoveries from dilute samples.

Vivacon 500 are centrifugal devices for DNA concentration and buffer exchange applications for samples up to 500μl. They are also the product of choice for protein fractionation applications in this volume range. For optimal performance with very diluted samples, e.g. forensic specimens, Vivacon 500 is equipped with patented Hydrosart™ regenerated cellulose membranes with extremely low unspecific binding properties and high flux. High recoveries and excellent reproducibility are paired with convenience offered by volume graduation and molecular weight cut-offs printed on individual devices. Vivacon 500 features a horizontal membrane design optimal for DNA fractionation (primer removal) as well as protein fractionation (peptide removal). The possibility of a reverse spin after sample processing assures complete concentrate recovery which is especially important when working with low sample concentrations.

  • Convenient handling in fixed-angle rotors
  • Low binding material
  • Reverse spin possibility
  • MWCO printed on
  • Complete and highly reproducible concentrate recovery
TRUSTED_SUSTAINABILITY

Specifications

Concentration 50ng/mL
Concentration Time 18 min.
Filter Type Ultrafiltration
For Use With (Equipment) Centrifuge
Material Polycarbonate
Material (Membrane) Regenerated Cellulose
MWCO 50 kDa
Sample Volume 500 μL
Sterility Non-sterile
Filtration Area 0.32 cm2
For Use With (Application) DNA concentration, desalting, buffer exchange, fractionation
Max. Force 5000xG
Quantity 100 Pack
Sample Type DNA, peptide
Type Concentrator
Volume (Metric) 0.5 mL
Volume (Metric) Hold Up <5 μL
Dimensions (L x W) 1.77 x 0.48 in. (4.5 x 1.24cm)
Length (English) 1.77 in.
Length (English) Back Spin 1.87 in.
Length (Metric) 4.5 cm
Length (Metric) Back Spin 4.75 cm
Width (English) 0.48 in.
Width (Metric) 1.24 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.