Learn More
Description
For up to 500 μL, Vivacon™ 500 are centrifugal units for fast DNA concentration and buffer exchange of 500 μL samples. They are also the product of choice for protein fractionation applications. The use of Hydrosart™ membranes results in high recoveries from dilute samples.
Vivacon 500 are centrifugal devices for DNA concentration and buffer exchange applications for samples up to 500μl. They are also the product of choice for protein fractionation applications in this volume range. For optimal performance with very diluted samples, e.g. forensic specimens, Vivacon 500 is equipped with patented Hydrosart™ regenerated cellulose membranes with extremely low unspecific binding properties and high flux. High recoveries and excellent reproducibility are paired with convenience offered by volume graduation and molecular weight cut-offs printed on individual devices. Vivacon 500 features a horizontal membrane design optimal for DNA fractionation (primer removal) as well as protein fractionation (peptide removal). The possibility of a reverse spin after sample processing assures complete concentrate recovery which is especially important when working with low sample concentrations.
- Convenient handling in fixed-angle rotors
- Low binding material
- Reverse spin possibility
- MWCO printed on
- Complete and highly reproducible concentrate recovery
Specifications
Specifications
| Concentration | 50ng/mL |
| Concentration Time | 10 min. |
| Filter Type | Ultrafiltration |
| For Use With (Equipment) | Centrifuge |
| Material | Polycarbonate |
| Material (Membrane) | Regenerated Cellulose |
| MWCO | 100 kDa |
| Sample Volume | 500 μL |
| Sterility | Non-sterile |
| Filtration Area | 0.32 cm2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.