Learn More
Description
- Optimally suited for cell culture supernatant concentration, virus concentration and water concentration
- Utilizes a 200cm2 PES or Hydrosart membrane for high flow rate
- Thin-channel, flip-flow recirculation path provides high crossflow velocities with minimum pump speed
- Fast concentration using one or two cassettes
- A single module typically concentrates 1L by a factor of 50 in less than 30 minutes
- Two modules used in parallel concentrates 5L by a factor of 50 in under 75 minutes
- Near total recovery of the concentrate is achieved with a single rinse
- A 500ml sealed diafiltration reservoir can be added for rapid and convenient buffer exchange
- One unit comes with all necessary accessories (except pump and pump head)
- Includes tubing, pressure indicator, flow restrictor and high pressure pump tubing
- Required equipment: peristaltic pump with a size 16 pump head
- Can be easily cleaned and re-used several times
- Clear housing for process monitoring
- Ready-to-use, disposable cassettes
- Easily set up two cassettes to run simultaneously for faster concentration
- Use a pre-assembled Y connector with the correct tubing sizes and fittings
- Size 16 tubing is used for connecting the two Vivaflow 200 units
- Size 15 tubing to connect to the pump, with a size 15 pump head
Specifications
Specifications
| Filter Type | Ultrafiltration |
| Format | Cassette |
| For Use With (Equipment) | Labscale TFF System |
| Material | Acrylic |
| Material (Membrane) | Cellulose |
| MWCO | 10 kDa |
| Sample Volume | Up to 2.5 L |
| Sterility | Non-sterile |
| Filtration Area | 200 cm2 |
| For Use With (Application) | Cell culture supernatant concentration, virus concentration and water concentration |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.