Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Vivaflow™ 200 Crossflow Cassettes

Catalog No. 14558319 Shop All Sartorius Products
Change view
Click to view available options
Material (Membrane):
Cellulose
PES
MWCO:
0.2 μm
10 kDa
100 kDa
30 kDa
5 kDa
50 kDa
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material (Membrane) MWCO
14-558-319 PES 5 kDa
14-558-320 PES 30 kDa
14-558-318 PES 10 kDa
14-558-322 PES 100 kDa
14-558-315 Cellulose 10 kDa
14-558-321 PES 50 kDa
14-558-317 Cellulose 30 kDa
14-558-323 PES 0.2 μm
14-558-314 Cellulose 100 kDa
14-558-316 Cellulose 5 kDa
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 14-558-319 Supplier Sartorius Supplier No. VF20P1
Add to Cart
Add to Cart

Offers a standard for ease of use, reliability and flexibility unrivalled by other filtration systems. Sartorius Vivaflow 200 Crossflow Cassettes are ready to use, reusable laboratory crossflow cassettes for concentrating up to 5L samples. Available in different MWCOs and membrane types for different application needs.

Rapid, Efficient Concentration
  • Optimally suited for cell culture supernatant concentration, virus concentration and water concentration
  • Utilizes a 200cm2 PES or Hydrosart membrane for high flow rate
  • Thin-channel, flip-flow recirculation path provides high crossflow velocities with minimum pump speed
  • Fast concentration using one or two cassettes
    • A single module typically concentrates 1L by a factor of 50 in less than 30 minutes
    • Two modules used in parallel concentrates 5L by a factor of 50 in under 75 minutes
  • Near total recovery of the concentrate is achieved with a single rinse
  • A 500ml sealed diafiltration reservoir can be added for rapid and convenient buffer exchange
Convenient and Easy to Use
  • One unit comes with all necessary accessories (except pump and pump head)
    • Includes tubing, pressure indicator, flow restrictor and high pressure pump tubing
    • Required equipment: peristaltic pump with a size 16 pump head
  • Can be easily cleaned and re-used several times
  • Clear housing for process monitoring
  • Ready-to-use, disposable cassettes
  • Easily set up two cassettes to run simultaneously for faster concentration
    • Use a pre-assembled Y connector with the correct tubing sizes and fittings
    • Size 16 tubing is used for connecting the two Vivaflow 200 units
    • Size 15 tubing to connect to the pump, with a size 15 pump head
TRUSTED_SUSTAINABILITY

Specifications

Filter Type Ultrafiltration
Format Cassette
For Use With (Equipment) Labscale TFF System
Material Acrylic
Material (Membrane) PES
MWCO 5 kDa
Sample Volume Up to 2.5 L
Sterility Non-sterile
Filtration Area 200 cm2
For Use With (Application) Cell culture supernatant concentration, virus concentration and water concentration
Includes Pressure indicator, flow restrictor and size 16 PVC peristaltic tubing and fittings
Max. Pressure 60psi
Pump Flow Rate 200 to 400mL/min.
Quantity 1 Each
Temperature (English) Max. 140°F
Temperature (Metric) Max 60°C
Type Crossflow Cassette
Volume (Metric) 5 L
Volume (Metric) Hold Up 5.3 mL
Dimensions (L x W x H) 4.96 x 1.49 x 5.43 in. (12.6 x 3.8 x 13.8cm)
Length (Metric) 12.6 cm
Width (Metric) 3.8 cm
Width (Metric) Channel 1 cm
Height (Metric) 13.8 cm
Height (Metric) Channel 0.04 cm
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.