Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Vivaflow™ 50 Crossflow Casettes

Catalog No. 14558306 Shop All Sartorius Products
Change view
Click to view available options
Material (Membrane):
PES
MWCO:
10 kDa
100 kDa
3 kDa
30 kDa
50 kDa
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material (Membrane) MWCO
14-558-306 PES 10 kDa
14-558-312 PES 3 kDa
14-558-310 PES 100 kDa
14-558-308 PES 30 kDa
14-558-309 PES 50 kDa
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 14-558-306 Supplier Sartorius Supplier No. VF05P0

Single use. Sartorius Vivaflow™ 50 Crossflow Casettes are ready-to-use for filtration and concentration of samples from 100 mL to 5L.

Optimally suited for applications like cell culture supernatant concentration, virus concentration and water concentration e.g. in fish breading facilities, this economical product offers a standard of ease of use, reliability and flexibility unrivalled by any laboratory filtration system in the market. The thin-channel, flip-flow recirculation path of the modules provides high crossflow velocities with minimum pump speed requirements. A single 50cm2 module typically reduces 500 ml to 15 ml in less than 50 min. Nearly total recovery of the concentrate is achieved with a single rinse. Up to six of the unique interlocking modules can be connected for easy scale up resulting in a capacity of several liters. Vivaflow 50 is intended for single use. All accessories for sample concentration, except a peristaltic pump with a pump head for size 16 tubing, are included in the package. A 500 ml sealed diafiltration reservoir can be added to the system for rapid and convenient buffer exchange.

  • High flow rate and recovery
  • Clear housing for process monitoring
  • Ready to use single cassettes
  • Easy set up of multiple cassettes
  • Economical and disposable product

Equipment required

The standard Vivaflow 50 package comes with two cassettes, tubing, flow restrictor, one series interconnector and high pressure pump tubing. All you need is a peristaltic pump with a pump head capable of handling 6.4mm OD tubing (size 16).

Running more than two Vivaflow 50 devices in parallel and series

For speeding up the concentration, as many as six Vivaflow 50 cassettes can easily be connected using 2 T-connectors for connecting 2 Vivaflow cassettes at the inlet and at the outlet and series interconnectors for connecting the devices in between. An optional pressure indicator (Fisher Cat# 14-558-332) may be used at the inlet for optimal control.

Unique in its performance

A single 50cm2 module will typically reduce 500mL to less than 15ml in under 50 minutes. Less than 10mL minimum system recirculation for highest concentrations. Less than 1.5mL non recoverable hold up volume. Near total recoveries achievable with a single 10mL rinse.

TRUSTED_SUSTAINABILITY

Specifications

Filter Type Ultrafiltration
Format Cassette
For Use With (Equipment) Labscale TFF System
Material Polycarbonate
Material (Membrane) PES
MWCO 10 kDa
Sample Volume 100 mL to 3 L
Sterility Non-sterile
Filtration Area 50 cm2
For Use With (Application) Cell culture supernatant concentration, virus concentration and water concentration
Includes Filtrate tube, size 16 peristaltic tubing, flow restrictor and fittings
Max. Pressure 45psi
Pump Flow Rate 200 to 400mL/min.
Quantity 2 Pack
Temperature (English) Max. 140°F
Temperature (Metric) Max 60°C
Type Crossflow Cassette
Volume (English) 0.33 to 33.8 oz.
Volume (Metric) 1 L
Dimensions (L x W x H) 4.21 x 0.98 x 3.3 in. (10.7 x 2.5 x 8.4cm)
Length (English) 4.21 in.
Length (Metric) 10.7 cm
Width (English) 0.98 in.
Width (English) Channel 0.59 in.
Width (Metric) 2.5 cm
Width (Metric) Channel 1.5 cm
Height (English) 3.3 in.
Height (English) Channel 0.011 in.
Height (Metric) 8.4 cm
Height (Metric) Channel 0.03 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.