Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Vivaspin™ 20 Centrifugal Concentrator
GSA_VA

Catalog No. 14558503 Shop All Sartorius Products
Change view
Click to view available options
MWCO:
0.2 μm
10 kDa
100 kDa
1000 kDa
3 kDa
30 kDa
300 kDa
5 kDa
50 kDa
Quantity:
12/Pk.
48/Pk.
Pore Size:
0.2 μm
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. MWCO Quantity Pore Size
14-558-503 5 kDa 12/Pk. -
14-558-510 100 kDa 48/Pk. -
14-558-505 30 kDa 12/Pk. -
14-558-516 0.2 μm 48/Pk. 0.2 μm
14-558-506 30 kDa 48/Pk. -
14-558-508 50 kDa 48/Pk. -
14-558-509 100 kDa 12/Pk. -
14-558-502 10 kDa 48/Pk. -
14-558-513 1000 kDa 12/Pk. -
14-558-504 5 kDa 48/Pk. -
14-558-512 300 kDa 48/Pk. -
14-558-507 50 kDa 12/Pk. -
14-558-501 10 kDa 12/Pk. -
14-558-517 3 kDa 12/Pk. -
14-558-511 300 kDa 12/Pk. -
14-558-518 3 kDa 48/Pk. -
14-558-515 0.2 μm 12/Pk. 0.2 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
Catalog No. 14-558-503 Supplier Sartorius Supplier No. VS2011
Add to Cart
Add to Cart

Offers increased volume flexibility and performance. VivaspinTM 20 Centrifugal Concentrator handles up to 20mL in swing bucket rotors and 14mL in fixed angle rotors accepting 50mL centrifuge tubes. Features twin vertical membranes for unparalleled filtration speed and >100-fold concentration.

Offers increased volume flexibility and performance. VivaspinTM 20 Centrifugal Concentrator handles up to 20mL in swing bucket rotors and 14mL in fixed angle rotors accepting 50mL centrifuge tubes. Features twin vertical membranes for unparalleled filtration speed and >100-fold concentration.

Vivaspin™ 20 centrifugal concentrators have been developed to offer increased volume flexibility and performance. Vivaspin™ 20 handles up to 20mL in swing bucket rotors and 14mL in fixed angle rotors accepting 50mL centrifuge tubes. Vivaspin™ 20 features twin vertical membranes for unparalleled filtration speed and >100-fold concentration. Volume graduation allows to monitor the concentration process and the modified dead stop pocket simplifies pipetting of the final concentrate. For higher process flexibility, Vivaspin™ 20 is available with unique accessories and operating methods. For one-step desalting following concentration, a diafiltration cup filled with buffer is placed on the concentrator and spun to achieve 98% salt removal, instead of two spins to achieve the same result with the traditional refill and re-spin procedure. When an appropriate centrifuge is unavailable, or for single sample processing, for gas pressure filtration, Vivaspin™ 20 can be filled with up to 15ml and then pressurised for bench top concentration. For even faster processing, gas pressure filtration can be combined with centrifugation. Pressure-fugation is particularly useful for difficult or viscous samples such as serum, when using a low process temperature, or generally when minimum process time is essential.

  • High flux PES membrane
  • High retentate recovery > 90 %
  • Vertical twin membrane for ultra fast concentration
  • Universal rotor compatibility
  • Safe sample concentration due to dead-stop volume
  • Convenient concentrate recovery by pipetting
  • Volume graduation for easy reading/monitoring
  • 100-fold concentration with twin vertical membranes
  • Additional option for pressurized operation

Rotor Compatibility

Please note: Vivaspin™ 20 (30mm x 116mm) is designed to fit into rotors that can accommodate Falcon 50 ml conical bottom tubes, e.g. Beckman Allegra 25R with TS-5.1-500 swing-out rotor with BUC 5 buckets and 368327 adaptors; Beckman TA-10.250 25° fixed angle rotor with 356966 adaptors; Multifuge 3 S-R with 75006445 swing out rotor with 75006441 buckets and adaptors for Falcon 50 ml conical bottom tubes. These devices are not designed to fit into rotors that only accept round bottom 29mm x 105mm tubes, e.g. Sorvall SS34 or Beckmann JA 20. If your rotor accepts only 29mm x 105mm round bottom tubes, please use the Vivaspin™ 15, which can be used in either round bottom or conical centrifuge tubes

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Disposable Single-use
Filter Type Ultrafiltration
For Use With (Equipment) Centrifuge
Material (Membrane) PES
MWCO 5 kDa
Sample Volume 20 mL
Sterility Non-sterile
Quantity 12/Pk.
Type Centrifugal Concentrator
Volume (Metric) 14 mL
Length (Metric) 116 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.