Learn More
Description
Up to 30-fold concentration of the sample can be achieved with recovery of the target molecule typically exceeding 95%. Patented dead-stop technology ensures that samples cannot be concentrated to dryness and enables direct concentrate recovery.
- Accept sample volumes from 100μLto 20mL, with a range of molecular weight cutoff values from Mr3000 to 100,000
- Vertical polyethersulfone semi-permeable membrane separating upper and lower chamber resists blockage and tolerates high flow rate
- Centrifugation forces solvent through membrane, leaving concentrated sample in the upper chamber
- Compatible pH range: 1 to 9
- Active membrane area: 0.5cm2
For maximum recovery, select a MWCO at least 50% smaller than the molecular size of the species of interest.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Material (Membrane) | PES |
| MWCO | 3 kDa |
| Filtration Area | 0.5 cm2 |
| Quantity | 25/Pk. |
| Volume (Metric) | 500 μL |
| Length (Metric) | 50 mm |
| Width (Metric) | 11 mm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
