Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Vivaspin™ 6 Sample Concentrators

Catalog No. 45001575 Shop All Cytiva Products
Change view
Click to view available options
MWCO:
10 kDa
100 kDa
3 kDa
30 kDa
5 kDa
50 kDa
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. MWCO
45-001-575 50 kDa
45-001-574 30 kDa
45-001-572 5 kDa
45-001-573 10 kDa
45-001-576 100 kDa
45-001-571 3 kDa
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 45-001-575 Supplier Cytiva Supplier No. 28932318
Add to Cart
Add to Cart

Use for one-step, non-denaturing sample concentration in a single tube for minimal sample handling and reduced sample loss. Cytiva Life Vivaspin™ 6 Sample Concentrators cater to scientists working with sample volumes ranging from 100 μl to 20 ml, with a range of MWCO values from Mr 3 000 to 100 000.

Up to 30-fold concentration of the sample can be achieved with recovery of the target molecule typically exceeding 95%. Patented dead-stop technology ensures that samples cannot be concentrated to dryness and enables direct concentrate recovery.

  • Accept sample volumes from 100μLto 20mL, with a range of molecular weight cutoff values from Mr3000 to 100,000
  • Vertical polyethersulfone semi-permeable membrane separating upper and lower chamber resists blockage and tolerates high flow rate
  • Centrifugation forces solvent through membrane, leaving concentrated sample in the upper chamber
  • Compatible pH range: 1 to 9
  • Active membrane area: 2.5cm2

For maximum recovery, select a MWCO at least 50% smaller than the molecular size of the species of interest.

TRUSTED_SUSTAINABILITY

Specifications

Material (Membrane) PES
MWCO 50 kDa
Filtration Area 2.5 cm2
Quantity 25/Pk.
Volume (Metric) 6 mL
Length (Metric) 122 mm
Width (Metric) 17 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.