Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Vivaspin™ Turbo 4 Ultrafiltration Unit

Catalog No. 14558703 Shop All Sartorius Products
Change view
Click to view available options
MWCO:
10 kDa
100 kDa
3 kDa
30 kDa
5 kDa
50 kDa
Quantity:
100/Pk.
25/Pk.
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. MWCO Quantity
14-558-703 3 kDa 25/Pk.
14-558-704 3 kDa 100/Pk.
14-558-705 5 kDa 25/Pk.
14-558-706 5 kDa 100/Pk.
14-558-707 10 kDa 25/Pk.
14-558-708 10 kDa 100/Pk.
14-558-709 30 kDa 25/Pk.
14-558-710 30 kDa 100/Pk.
14-558-711 50 kDa 25/Pk.
14-558-712 50 kDa 100/Pk.
14-558-713 100 kDa 25/Pk.
14-558-714 100 kDa 100/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 14-558-703 Supplier Sartorius Supplier No. VS04T91
Add to Cart
Add to Cart

Handles up to 4mL sample volume in 4 swing-out and fixed angle rotors accepting 15mL centrifuge tubes. Sartorius Vivaspin Turbo 4 Ultrafiltration Unit, features twin vertical membranes and sleek internal design for fastest sample concentration with highest recoveries.

  • High flux PES membrane
  • Retentate recovery >95% x pH 1-14 suited polystyrene housing with high chemical compatibility
  • Universal rotor compatibility
  • Volume graduation for easy detection / monitoring of the concentrate volume
  • UV joining technology allows for smooth joint transition between membrane and plastic housing, allowing collection of complete concentrated sample into unique pipette friendly dead stop pocket
TRUSTED_SUSTAINABILITY

Specifications

Description Turbo 4 Ultrafiltration Unit
Disposable Single-use
Filter Type Ultrafiltration
For Use With (Equipment) Centrifuge
Material (Membrane) PES
MWCO 3 kDa
Sample Volume 4 mL
Sterility Non-sterile
For Use With (Application) Concentration, Desalting, buffer exchange
Max. Force Swing-out rotor : 4000 x gAngle rotor : 7500 x g
Quantity 25/Pk.
Sample Type Sample type: Nanoparticles; Peptide; Protein
Type Centrifugal Concentrator
Volume (Metric) 4 mL
Length (Metric) 1225 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.