Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Viviparous 8 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986257
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6257 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6257 Supplier Bioss Supplier No. BS4993R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Involved in the regulation of meristem development and seed maturation processes. Mediates regulation of embryonic regulatory genes and genes controlling abscisic acid (ABA) biosynthesis and turnover in developing seeds. May be required for the synthesis of small signaling molecules that integrates meristem and embryo formation in seeds.

Specifications

Antigen Viviparous 8
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from Viviparous 8.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.