Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Viz-Up™ ANSI Type R Class 2 Expandable Mesh Breakaway Vest

Catalog No. 19046483 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
One Size
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-046-483 One Size
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-046-483 Supplier PIP™ Supplier No. 47200/O
Only null left
Add to Cart
Add to Cart

Type R/Class 2 apparel is necessary for workers exposed to traffic traveling over 25 mph and those who work against complex backgrounds. Ideal for construction, municipalities, shipyards, or anywhere that hi-vis apparel is required.

  • Breathable polyester mesh
  • Breakaway provides additional safety around extreme traffic hazards, or moving machinery and equipment
  • Adjustable hook & loop sides for universal fit
  • Hook & Loop closure
  • 2 in. silver reflective tape
  • 1 interior chest pocket
  • 2 interior pockets

Specifications

Product Type High-visibility Vest
Certifications/Compliance ANSI/ISEA 107-2015 Class 3 Type R
Closure Type Hook and Loop
Construction 5 Point Breakaway/Expandable
Color Hi-vis Orange
Size One Size
Material Polyester Mesh
Length (English) 28 in.
Length (Metric) 71 cm
Size (English) Chest 26.5 in.
No. of Pockets 3
Size (Metric) Chest 67.3 cm
Tape Type Level 2 Silver Reflective (2 in.)
For Use With (Application) Construction, Municipalities, Shipyard, and Railroad Workers
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.