Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VKORC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534358
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534358 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534358 Supplier Thermo Scientific Supplier No. PA534358
Add to Cart
Add to Cart

VKORC1 Polyclonal Antibody

VKORC1 Polyclonal Antibody for Western Blot, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen VKORC1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene VKORC1
Gene Accession No. Q9BQB6, Q9CRC0
Gene Alias D7Wsu86e; EDTP308; IMAGE3455200; MST134; MST576; MSTP134; MSTP576; phylloquinone epoxide reductase; UNQ308/PRO351; vitamin K dependent clotting factors deficiency 2; vitamin K epoxide reductase complex subunit 1; vitamin K epoxide reductase complex, subunit 1; vitamin K1 2,3-epoxide reductase subunit 1; vitamin K1 epoxide reductase (warfarin-sensitive); VKCFD2; VKOR; Vkorc1
Gene Symbols VKORC1
Host Species Rabbit
Immunogen A 15 amino acid synthetic peptide near the amino terminus of human VKORC1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27973, 79001
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.