Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VKORC1L1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562881
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562881 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562881 Supplier Invitrogen™ Supplier No. PA562881
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LIINYKRLVY LNEAWKRQLQ PKQD Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

Involved in vitamin K metabolism. Can reduce inactive vitamin K 2,3-epoxide to active vitamin K (in vitro), and may contribute to vitamin K-mediated protection against oxidative stress. Plays a role in vitamin K-dependent gamma-carboxylation of Glu residues in target proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VKORC1L1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene VKORC1L1
Gene Accession No. Q8N0U8
Gene Alias 2310024K08Rik; vitamin K epoxide reductase complex subunit 1 like 1; vitamin K epoxide reductase complex subunit 1-like protein 1; vitamin K epoxide reductase complex, subunit 1-like 1; VKORC1L1; VKORC1-like protein 1
Gene Symbols VKORC1L1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human VKORC1L1. Recombinant protein control fragment (Product #RP-101592).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 154807
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.