Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VLDLR Antibody, Alexa Fluor™ 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299696AF488
Description
VLDLR Polyclonal specifically detects VLDLR in Human samples. It is validated for ELISA.Specifications
VLDLR | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
Alexa Fluor 488 | |
CARMQ1, CHRMQ1, FLJ35024, very low density lipoprotein receptor, very low-density lipoprotein receptor, VLDL receptor, VLDL-R, VLDLRCH | |
Produced in rabbits immunized with purified, recombinant VLDLR (NP_003374.3; Met 1-Ser 797). | |
0.1 mL | |
Apoptosis, Cholesterol Metabolism, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience | |
7436 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction