Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VLDLR Monoclonal Antibody (6A6)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA524790

Catalog No. PIMA524790


Only null left
Add to Cart

Description

Description

VLDLR Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

The low density lipoprotein receptor (LDLR) gene family consists of cell surface proteins involved in receptor-mediated endocytosis of specific ligands. This gene encodes a lipoprotein receptor that is a member of the LDLR family and plays important roles in VLDL-triglyceride metabolism and the reelin signaling pathway. Mutations in this gene cause VLDLR-associated cerebellar hypoplasia. Alternative splicing generates multiple transcript variants encoding distinct isoforms for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

VLDLR
Monoclonal
1 mg/mL
PBS with 0.02% sodium azide
P98155, P98156, P98166
VLDLR
Synthetic peptide corresponding to the C-terminus of human VLDL Receptor.
100 μL
Primary
Human, Mouse, Rat, Bovine
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
6A6
Unconjugated
VLDLR
AA408956; AI451093; AW047288; CAMRQ1; CARMQ1; CHRMQ1; FLJ35024; non O-glycosylated VLDL-R; very low density lipoprotein receptor; very low density lipoprotein receptor VLDL-R2; very low-density lipoprotein receptor; VLDL receptor; VLDLR; VLDL-R; VLDLRCH
Mouse
Protein G
RUO
22359, 25696, 282123, 7436
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.