Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VLDLR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116385
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116385 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116385 Supplier Invitrogen™ Supplier No. PA5116385
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

VLDLR Polyclonal Antibody for Western Blot

The low density lipoprotein receptor (LDLR) gene family consists of cell surface proteins involved in receptor-mediated endocytosis of specific ligands. This gene encodes a lipoprotein receptor that is a member of the LDLR family and plays important roles in VLDL-triglyceride metabolism and the reelin signaling pathway. Mutations in this gene cause VLDLR-associated cerebellar hypoplasia. Alternative splicing generates multiple transcript variants encoding distinct isoforms for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VLDLR
Applications Western Blot
Classification Polyclonal
Concentration 0.36 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene VLDLR
Gene Accession No. P98155
Gene Alias AA408956; AI451093; AW047288; CAMRQ1; CARMQ1; CHRMQ1; FLJ35024; non O-glycosylated VLDL-R; very low density lipoprotein receptor; very low density lipoprotein receptor VLDL-R2; very low-density lipoprotein receptor; VLDL receptor; VLDLR; VLDL-R; VLDLRCH
Gene Symbols VLDLR
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 608-642 amino acids from human VLDLR.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7436
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.