Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ VLDLR, Rat, Unlabeled, Clone: 4C4G5,

Catalog No. MABS1288 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABS1288 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS1288 Supplier MilliporeSigma™ Supplier No. MABS1288
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Pulmonary surfactant-associated protein D (UniProt P35247; also known as Collectin-7, Lung surfactant protein D, PSP-D, SP-D) is encoded by the SFTPD (also known as COLEC7, PSPD, SFTP4) gene (Gene ID 6441) in human. Surfactant, produced by alveolar type II cells and airway Clara cells, resides at the air-liquid interface of the lungs and is most commonly known for its role in reducing surface tension. Surfactant is a lipoprotein complex composed of approximately 10% proteins, including surfactant protein A (SP-A), SP-B, SP-C, and SP-D. SP-A and SP-D belong to the collectin family of proteins and can modulate innate immunity. SP-D is known to enhance pulmonary clearance of pathogens, such as Pseudomonous aerginosa, Klebsiella pneumonia, respiratory syncytial virus (RSV) and Influenza virus. SP-D is also shown to modify allergic responsee in the lungs by binding common allergens, including house dust mite and pollen particles. SP-A-deficient and SP-D-deficient mice are susceptible to bleomycin-induced lung injury and display increased cellular inflammation, more severe lung fibrosis, and reduced survival. In addition to lung, SP-D is also reported to be detected in extrapulmonary tissues, such as kidney and pancreas. Serum levels of either SP-A or SP-D in human are correlated with mortality in patients with pulmonary fibrosis. Human SP-D is produced with a signal peptide sequence (a.a. 1-20), which is removed posttranslationally to yield the mature protein (a.a. 21-375) that contains a collagen-like domain (a.a. 46-222), a coiled coil region (a.a. 223-252), and a C-type lectin domain (a.a. 260-375).
TRUSTED_SUSTAINABILITY

Specifications

Antigen VLDLR
Applications Western Blot
Classification Monoclonal
Clone 4C4G5
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rat IgG2b in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene Vldlr
Gene Accession No. P98156
Host Species Rat
Immunogen GST-tagged recombinant mouse VLDLR cytoplasmic tail (Hirota, Y., et al. (2015). J. Comp. Neurol. 523(3):463-478).
Purification Method Protein G Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_001154892
Test Specificity Clone 4C4G5 (4G) detected VLDLR expression in embryonic and neonatal brain tissues from wild-type, but not Vldlr-knockout mice (Hirota, Y., et al. (2015). J. Comp. Neurol. 523(3):463-478).
Target Species Mouse
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.