Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VLK Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110916
Description
Immunogen sequence: TEYQCIPDST IPQEDYRCWP SYHHGSCLLS VFNLAEAVDV CESHAQCRAF VVTNQTTWTG RQLVFFKTGW SQVVPDPNKT T.
VLK was identified as a novel protein kinase that was induced after the differentiation of cultured embryonic stem cells into mesendoderm. It has no homologs in invertebrates, but is highly conserved in vertebrate species although it does not belong to any known protein kinase groups. VLK is initially expressed in E-cadherin-positive anterior visceral endoderm and mesendoderm, but its expression is later confined to E-cadherin-negative mesenchyme. It is enriched in the Golgi apparatus and is thought to regulate the rate of protein export from the Golgi. Targeted disruption of VLK in mice leads to a defect in lung development and neonatal lethality. It has been suggested that mutations in VLK may be associated with the allergic condition atopy.
Specifications
| VLK | |
| Polyclonal | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q504Y2 | |
| PKDCC | |
| Recombinant Protein corresonding to Human VLK. Recombinant protein control fragment (Product #RP-95072). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| PKDCC | |
| Adtk1; AI115348; AW548124; ESTM17; Extracellular tyrosine-protein kinase PKDCC; MAd1; Pkdcc; protein kinase domain containing, cytoplasmic; protein kinase domain containing, cytoplasmic homolog; protein kinase domain-containing protein, cytoplasmic; Protein kinase-like protein SgK493; RGD1311939; SGK493; Sugen kinase 493; vertebrate lonesome kinase; Vlk; X83346 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 91461 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction