Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VMAT1, Mouse anti-Mouse, Rat, Clone: N440/21, Millipore Sigma™

Catalog No. MABN254725 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN254725 25 μL
MABN2547100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABN254725 Supplier MilliporeSigma Supplier No. MABN254725UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-VMAT1, clone N440 21, Cat. No. MABN2547, is a highly specific mouse monoclonal antibody that targets Vesicular Monoamine Transporter 1 and has been tested for use in Western Blotting.

Chromaffin granule amine transporter (UniProt: Q8R090; also known as Solute carrier family 18 member 1, Vesicular amine transporter 1, VMAT1, VAT1) is encoded by the Scl19a1 (also known as Vmat1) gene (Gene ID: 110877) in murine species. Vesicular monoamine transporters (VMATs) are involved in presynaptic storage and release of neurotransmitters. They are located in the presynaptic region and are involved in the transport of monoamines into storage vesicle, which are released upon arrival of an action potential. Two structurally related, but pharmacologically distinct, VMATs have been described (VMAT1 and VMAT2) that are encoded by separate genes. VMAT1 is a multi-pass membrane acidic glycoprotein that is found in both large dense-core vesicles and in small synaptic vesicles. It s both C- and N-terminal regions are located in the cytosolic side of the vesicle. VMAT1 is detected in adrenal medulla, and brain (at protein level). In brain, specifically found in the medulla oblongata, pons, prefrontal cortex, striatum, dentate gyrus and hippocampus. VMAT1 is shown to play a key role in survival of hippocampal neurons and may contribute to neurocognitive deficits observed in neuropsychiatric disorders. (Ref.: Multani, PK., et al. (2013). Neurosci. 232; 32-44).
TRUSTED_SUSTAINABILITY

Specifications

Antigen VMAT1
Applications Western Blot
Classification Monoclonal
Clone N440/21
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1M Tris-Glycine, 0.15M NaCl, 0.05% sodium azide, pH7.4 with 0.05% sodium azide.
Gene Alias Chromaffin granule amine transporter;Solute carrier family 18 member 1;Vesicular amine transporter 1;VAT1
Gene Symbols Scl18a1;Vmat1
Host Species Mouse
Immunogen Recombinant fragment corresponding to 93 amino acids from the N-terminal first lumenal region of murine Vesicular amine transporter 1 (VMAT1).
Purification Method Protein G purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_694694.1
Test Specificity Clone N440/21 specifically detects Vesicular monoamine transporter 1 (VMAT1) and does not show reactivity with VMAT2. It targets an epitope with in 93 amino acids from the N-terminal first lumenal region.
Target Species Rat, Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.