Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VMAT2 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 9 publications
$243.50 - $499.50
Specifications
Antigen | VMAT2 |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500-1:1000, Immunohistochemistry 1:100-1:250, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen 1:100-1:250 |
Applications | Immunohistochemistry (Frozen), Immunofluorescence, Immunocytochemistry, Immunohistochemistry, Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB018004
![]() |
Novus Biologicals
NB11068123SS |
0.025 mg |
Each for $243.50
|
|
|||||
NB11068123
![]() |
Novus Biologicals
NB11068123 |
0.1 mg |
Each for $499.50
|
|
|||||
Description
VMAT2 Polyclonal specifically detects VMAT2 in Human, Mouse, Rat, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.Specifications
VMAT2 | |
Western Blot 1:500-1:1000, Immunohistochemistry 1:100-1:250, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen 1:100-1:250 | |
Polyclonal | |
Rabbit | |
Human, Rabbit, Mouse, Rat | |
Q05940, Q8BRU6, Q01827 | |
6571 | |
Reacts with an internal sequence of the human, mouse and rat VMAT2 protein. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
55.7 kDa |
1 mg/mL | |
Immunohistochemistry (Frozen), Immunofluorescence, Immunocytochemistry, Immunohistochemistry, Western Blot | |
Unconjugated | |
RUO | |
PBS with 0.02% Sodium Azide | |
MGC120477, monoamine neurotransmitter transporter, Monoamine transporter, solute carrier family 18 (vesicular monoamine), member 2, Solute carrier family 18 member 2, SVAT, SVMTMGC120478, synaptic vesicular amine transporter, VAT2MGC26538, vesicle monoamine transporter type 2, vesicle monoamine/H+ antiporter, Vesicular amine transporter 2, VMAT2 | |
SLC18A2 | |
IgG | |
Affinity Purified | |
VMAT2 |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title