Learn More
Invitrogen™ VMAT2 Monoclonal Antibody (OTI9E11)

Description
VMAT2 Monoclonal Antibody for Flow, ICC/IF, IHC (P)
Specifications
Specifications
| Antigen | VMAT2 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI9E11 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | SLC18A2 |
| Gene Accession No. | Q05940 |
| Gene Alias | 1110037L13Rik; 9330105E13; MGC120477; MGC120478; MGC26538; MNAT; monoamine neurotransmitter transporter; monoamine transporter; SLC18A2; solute carrier family 18 (vesicular monoamine transporter), member 2; solute carrier family 18 (vesicular monoamine), member 2; Solute carrier family 18 A2 (vesicular monoamine transporter 2); solute carrier family 18 member 2; solute carrier family 18 member A2; solute carrier family 18, member 2; Svat; SVMT; synaptic vesicular amine transporter; VAT2; vesicle monoamine transporter type 2; vesicle monoamine/H+ antiporter; vesicular amine transporter 2; vesicular monoamine transporter 2; VMAT2; VMAT-2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.