Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VMAT2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA127477
Description
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
VMAT2 is involved in the ATP-dependent vesicular transport of biogenic amine neurotransmitters. pumps cytosolic monoamines including dopamine, norepinephrine, serotonin, and histamine into synaptic vesicles. It is a requisite for vesicular amine storage prior to secretion via exocytosis.
Specifications
VMAT2 | |
Polyclonal | |
Unconjugated | |
SLC18A2 | |
1110037L13Rik; 9330105E13; MGC120477; MGC120478; MGC26538; MNAT; monoamine neurotransmitter transporter; monoamine transporter; SLC18A2; solute carrier family 18 (vesicular monoamine transporter), member 2; solute carrier family 18 (vesicular monoamine), member 2; Solute carrier family 18 A2 (vesicular monoamine transporter 2); solute carrier family 18 member 2; solute carrier family 18 member A2; solute carrier family 18, member 2; Svat; SVMT; synaptic vesicular amine transporter; VAT2; vesicle monoamine transporter type 2; vesicle monoamine/H+ antiporter; vesicular amine transporter 2; vesicular monoamine transporter 2; VMAT2; VMAT-2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
214084, 25549, 6571 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q01827, Q05940, Q8BRU6 | |
SLC18A2 | |
Purified VMAT protein. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction