Learn More
Invitrogen™ VMAT2 Polyclonal Antibody

Description
VMAT2 Polyclonal Antibody for ELISA, ICC/IF, Western Blot
Specifications
Specifications
| Antigen | VMAT2 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 |
| Gene | SLC18A2 |
| Gene Accession No. | Q05940, Q8BRU6 |
| Gene Alias | 1110037L13Rik; 9330105E13; MGC120477; MGC120478; MGC26538; MNAT; monoamine neurotransmitter transporter; monoamine transporter; SLC18A2; solute carrier family 18 (vesicular monoamine transporter), member 2; solute carrier family 18 (vesicular monoamine), member 2; Solute carrier family 18 A2 (vesicular monoamine transporter 2); solute carrier family 18 member 2; solute carrier family 18 member A2; solute carrier family 18, member 2; Svat; SVMT; synaptic vesicular amine transporter; VAT2; vesicle monoamine transporter type 2; vesicle monoamine/H+ antiporter; vesicular amine transporter 2; vesicular monoamine transporter 2; VMAT2; VMAT-2 |
| Gene Symbols | SLC18A2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.