Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VMO1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585226
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585226 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585226 Supplier Invitrogen™ Supplier No. PA585226
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Keep as concentrated solution. Predicted reactivity: Pig (84%), Rhesus Monkey (92%). Positive Control: Target recombinant protein. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

VMO1 (Vitelline Membrane Outer Layer 1 Homolog) is a Protein Coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VMO1
Applications Dot Blot, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene Vmo1
Gene Accession No. Q5SXG7, Q7Z5L0
Gene Alias ERGA6350; ERGA6350 antibody; Gm741; MGC125880 antibody; MGC125881 antibody; PRO21055; PRO21055 antibody; RGD1559861; UNQ6350/PRO21055; vitelline membrane outer layer 1 homolog; vitelline membrane outer layer 1 homolog (chicken); vitelline membrane outer layer 1 homolog (chicken) antibody; vitelline membrane outer layer protein 1 homolog; vitelline membrane outer layer protein 1 homolog antibody; VMO1; VMO1 antibody
Gene Symbols Vmo1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human VMO1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284013, 327956
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.