Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VMO1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144150
Description
VMO1 Polyclonal Antibody for ICC/IF, IHC (P), ELISA
VMO1 (Vitelline Membrane Outer Layer 1 Homolog) is a Protein Coding gene.
Specifications
VMO1 | |
Polyclonal | |
Unconjugated | |
Vmo1 | |
ERGA6350; ERGA6350 antibody; Gm741; MGC125880 antibody; MGC125881 antibody; PRO21055; PRO21055 antibody; RGD1559861; UNQ6350/PRO21055; vitelline membrane outer layer 1 homolog; vitelline membrane outer layer 1 homolog (chicken); vitelline membrane outer layer 1 homolog (chicken) antibody; vitelline membrane outer layer protein 1 homolog; vitelline membrane outer layer protein 1 homolog antibody; VMO1; VMO1 antibody | |
Rabbit | |
Protein G | |
RUO | |
284013 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
Q7Z5L0 | |
Vmo1 | |
Recombinant Human Vitelline membrane outer layer protein 1 homolog protein (25-202AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction