Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VMP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578588
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578588 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578588 Supplier Invitrogen™ Supplier No. PA578588
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: Unboiled HepG2 membrane extract Predicted Reactivity: Mouse (96%), Rat (98%), Zebrafish (82%), Xenopus laevis (88%), Rhesus Monkey (99%), Chimpanzee (100%), Bovine (98%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

VMP1 is a stress-induced protein that, when overexpressed, promotes formation of intracellular vacuoles followed by cell death. May be involved in the cytoplasmic vacuolization of acinar cells during the early stage of acute pancreatitis.VMP1 is a transmembrane protein that co-localizes with LC3, a marker of the autophagosomes. It interacts with beclin-1, a mammalian autophagy initiator, through the VMP1-Atg domain, which is essential for autophagosome formation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VMP1
Applications Western Blot
Classification Polyclonal
Concentration 1.64 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene VMP1
Gene Accession No. Q96GC9
Gene Alias 3110098I04Rik; 4930579A11Rik; AI787464; EPG3; HSPC292; MIR21; mir-21a; NF-E2 induceble protein-2; NF-E2-inducible protein 2; ni-2; Protein ni-2; Tango5; TDC1; Tmem49; transmembrane protein 49; Vacuole membrane protein 1; VMP1; wu:fb05d08; wu:fb92g08; zgc:77153
Gene Symbols VMP1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the Extracellular domain of human TMEM49. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81671
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.