Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VN1R1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599936
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599936 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599936 Supplier Invitrogen™ Supplier No. PA599936
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total VN1R1.

Pheromones are chemical signals that elicit specific behavioral responses and physiologic alterations in recipients of the same species. The protein encoded by this gene is similar to pheromone receptors and is primarily localized to the olfactory mucosa. An alternate splice variant of this gene is thought to exist, but its full length nature has not been determined. Putative pheromone receptor. Expressed in the olfactory mucosa, very low expression in brain, lung and kidney. The chimpanzee and orangutan orthologous proteins do not exist, their genes are pseudogenes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VN1R1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene VN1R1
Gene Accession No. Q9GZP7
Gene Alias G-protein coupled receptor GPCR24; hGPCR24; V1RL1; V1R-like 1; V1r-like receptor 1; V3r-related gene protein; VN1R1; VNR19I1; vomeronasal 1 receptor 1; vomeronasal olfactory receptor chromosome 19 subtype I member 1; vomeronasal olfactory receptor, (chromosome 19) subtype I, member 1; vomeronasal type-1 receptor 1; ZVNH1; ZVNR1
Gene Symbols VN1R1
Host Species Rabbit
Immunogen A synthesized peptide derived from human VN1R1(Accession Q9GZP7), corresponding to amino acid residues G3-L53.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57191
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.