Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VN1R1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA534309
Description
Percent identity with other species by BLAST analysis: Human (100%) Chimpanzee (94%) Gorilla (88%) Orangutan (81%).
Pheromones are chemical signals that elicit specific behavioral responses and physiologic alterations in recipients of the same species. The protein encoded by this gene is similar to pheromone receptors and is primarily localized to the olfactory mucosa. An alternate splice variant of this gene is thought to exist, but its full length nature has not been determined. Putative pheromone receptor. Expressed in the olfactory mucosa, very low expression in brain, lung and kidney. The chimpanzee and orangutan orthologous proteins do not exist, their genes are pseudogenes.
Specifications
| VN1R1 | |
| Polyclonal | |
| Unconjugated | |
| VN1R1 | |
| G-protein coupled receptor GPCR24; hGPCR24; V1RL1; V1R-like 1; V1r-like receptor 1; V3r-related gene protein; VN1R1; VNR19I1; vomeronasal 1 receptor 1; vomeronasal olfactory receptor chromosome 19 subtype I member 1; vomeronasal olfactory receptor, (chromosome 19) subtype I, member 1; vomeronasal type-1 receptor 1; ZVNH1; ZVNR1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 57191 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q9GZP7 | |
| VN1R1 | |
| Synthetic 16 amino acid peptide from 3rd extracellular domain of human VN1R1. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction