Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VN1R1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534310
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534310 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534310 Supplier Thermo Scientific Supplier No. PA534310
Add to Cart
Add to Cart

VN1R1 Polyclonal Antibody

VN1R1 Polyclonal Antibody for IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen VN1R1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene VN1R1
Gene Accession No. Q9GZP7
Gene Alias G-protein coupled receptor GPCR24; hGPCR24; V1RL1; V1R-like 1; V1r-like receptor 1; V3r-related gene protein; VN1R1; VNR19I1; vomeronasal 1 receptor 1; vomeronasal olfactory receptor chromosome 19 subtype I member 1; vomeronasal olfactory receptor, (chromosome 19) subtype I, member 1; vomeronasal type-1 receptor 1; ZVNH1; ZVNR1
Gene Symbols VN1R1
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from C-terminus of human VN1R1.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57191
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.