Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VN1R2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113250
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113250 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113250 Supplier Invitrogen™ Supplier No. PA5113250
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

VN1R2 Polyclonal Antibody for Western Blot, ICC/IF, ELISA

Putative pheromone receptor. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen VN1R2
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene VN1R2
Gene Accession No. Q8NFZ6
Gene Alias G-protein coupled receptor GPCR25; hGPCR25; pheromone receptor; V1RL2; V1R-like 2; V1r-like receptor 2; VN1R2; vomeronasal 1 receptor 2; vomeronasal type-1 receptor 2
Gene Symbols VN1R2
Host Species Rabbit
Immunogen Recombinant Human Vomeronasal type-1 receptor 2 protein (115-170AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 317701
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.