Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VN1R4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102262
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102262 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102262 Supplier Invitrogen™ Supplier No. PA5102262
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total VN1R4.

Putative pheromone receptor. Various VN1R4 alleles are known. T The chimpanzee and gorilla orthologous proteins do not exist, their genes are pseudogenes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VN1R4
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene VN1R4
Gene Accession No. Q7Z5H5
Gene Alias CTD-2245F17.9; G-protein coupled receptor GPCR27; hGPCR27; pheromone receptor; V1RL4; V1R-like 4; V1r-like receptor 4; VN1R4; vomeronasal 1 receptor 4; vomeronasal type-1 receptor 4
Gene Symbols VN1R4
Host Species Rabbit
Immunogen A synthesized peptide derived from human VN1R4(Accession Q7Z5H5), corresponding to amino acid residues N142-L192.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 317703
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.