Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VNN1, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA564154
Description
Immunogen sequence: KLLLSQLDSH PSHSAVVNWT SYASSIEALS SGNKEFKGTV FFDEFTFVKL TGVAGNYTVC QKHighest antigen sequence identity to the following orthologs: Mouse - 65%, Rat - 71%.
Specifications
VNN1 | |
Polyclonal | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O95497 | |
VNN1 | |
Recombinant protein corresponding to Human VNN1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry | |
Unconjugated | |
VNN1 | |
HDLCQ8, Tiff66, pantetheinase, pantetheine hydrolase, vanin-1, vannin 1, vascular non-inflammatory molecule 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8876 | |
Store at 4°C short term. Store at −20°C long term. Avoid freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction