Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VNN2 Antibody (018), DyLight 755 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289970IR
Description
VNN2 Monoclonal antibody specifically detects VNN2 in Human samples. It is validated for ELISASpecifications
VNN2 | |
Monoclonal | |
DyLight 755 | |
EC 3.5.1, EC 3.5.1.92, FOAP-4, Glycosylphosphatidyl inositol-anchored protein GPI-80, GPI-80, GPI-80 variant protein 1, GPI-80 variant protein 2, GPI-80 variant protein 3, GPI-80 variant protein 4, pantetheinase, Protein FOAP-4, vanin 2, vanin-2, Vannin 2, vascular non-inflammatory molecule 2 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human VNN2 (NP_004656.2; Met 1-Ser 492). | |
0.1 mL | |
Endocrinology, Signal Transduction, Stem Cells | |
8875 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
018 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction