Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VNN2 Antibody (3F8F8H6), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP306441V
Description
VNN2 Monoclonal antibody specifically detects VNN2 in Human samples. It is validated for Flow Cytometry, ELISA, ELISASpecifications
| VNN2 | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA | |
| 3F8F8H6 | |
| Flow Cytometry, ELISA, Sandwich ELISA | |
| EC 3.5.1, EC 3.5.1.92, FOAP-4, Glycosylphosphatidyl inositol-anchored protein GPI-80, GPI-80, GPI-80 variant protein 1, GPI-80 variant protein 2, GPI-80 variant protein 3, GPI-80 variant protein 4, pantetheinase, Protein FOAP-4, vanin 2, vanin-2, Vannin 2, vascular non-inflammatory molecule 2 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human VNN2 (NP_004656.2; Met 1-Ser 492). | |
| 0.1 mL | |
| Endocrinology, Signal Transduction, Stem Cells | |
| 8875 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction