Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ VO-OHpic
GREENER_CHOICESDP

Catalog No. 359110 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
35-911-0 10 mg
35-915-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 35-911-0 Supplier Tocris Bioscience™ Supplier No. 3591/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent PTEN inhibitor

VO-OHpic is a potent inhibitor of PTEN (phosphatase and tensin homolog) (IC50 = 35 nM). Displays selectivity for PTEN over cysteine-based phosphatases such as SopB, MTM, PTPβ and SAC (IC50 values are 588 nM, 4.03, 57.5 and >10 μM respectively). Increases PIP2 and PIP3 levels; also increases Akt translocation in NIH/3T3 fibroblasts. Blocks the immunosuppressant activity of IDO-activated Tregs. Promotes antitumor inflammatory response in combination with immunotherapy and enhances anticancer chemotherapy in tumor-bearing mice.

Specifications

Quantity 10 mg
Product Type VO-OHpic
Purity >98%
Molecular Weight (g/mol) 361.16
Inhibitors VO-OHpic

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.