Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Volatile Organics Mix, Low Level, SPEX CertiPrep™
Shop All Spex CertiPrep Products

Click to view available options
Quantity:
1 mL
Description
Includes
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
| Analyte | CAS No. | Concentration |
|---|---|---|
| 1,1,1,2-Tetrachloroethane | 630-20-6 | 200μg/mL |
| 1,1,1-Trichloroethane | 71-55-6 | 200μg/mL |
| 1,1,2,2-Tetrachloroethane | 79-34-5 | 200μg/mL |
| 1,1,2-Trichloroethane | 79-00-5 | 200μg/mL |
| 1,1-Dichloroethane | 75-34-3 | 200μg/mL |
| 1,1-Dichloroethene | 75-35-4 | 200μg/mL |
| 1,1-Dichloropropene | 563-58-6 | 200μg/mL |
| 1,2,3-Trichlorobenzene | 87-61-6 | 200μg/mL |
| 1,2,3-Trichloropropane | 96-18-4 | 200μg/mL |
| 1,2,4-Trichlorobenzene | 120-82-1 | 200μg/mL |
| 1,2,4-Trimethylbenzene | 95-63-6 | 200μg/mL |
| 1,2-Dibromo-3-chloropropane | 96-12-8 | 200μg/mL |
| 1,2-Dibromoethane | 106-93-4 | 200μg/mL |
| 1,2-Dichlorobenzene | 95-50-1 | 200μg/mL |
| 1,2-Dichloroethane | 107-06-2 | 200μg/mL |
| 1,2-Dichloropropane | 78-87-5 | 200μg/mL |
| 1,3,5-Trimethylbenzene | 108-67-8 | 200μg/mL |
| 1,3-Dichlorobenzene | 541-73-1 | 200μg/mL |
| 1,3-Dichloropropane | 142-28-9 | 200μg/mL |
| 1,4-Dichlorobenzene | 106-46-7 | 200μg/mL |
| 2,2-Dichloropropane | 594-20-7 | 200μg/mL |
| 2-Chlorotoluene | 95-49-8 | 200μg/mL |
| 4-Chlorotoluene | 106-43-4 | 200μg/mL |
| Benzene | 71-43-2 | 200μg/mL |
| Bromobenzene | 108-86-1 | 200μg/mL |
| Bromochloromethane | 74-97-5 | 200μg/mL |
| Bromodichloromethane | 75-27-4 | 200μg/mL |
| Bromoform | 75-25-2 | 200μg/mL |
| Bromomethane | 74-83-9 | 200μg/mL |
| Carbon tetrachloride | 56-23-5 | 200μg/mL |
| Chlorobenzene | 108-90-7 | 200μg/mL |
| Chloroethane | 75-00-3 | 200μg/mL |
| Chloroform | 67-66-3 | 200μg/mL |
| Chloromethane | 74-87-3 | 200μg/mL |
| cis-1,2-Dichloroethene | 156-59-2 | 200μg/mL |
| cis-1,3-Dichloropropene | 10061-01-5 | 200μg/mL |
| Dibromochloromethane | 124-48-1 | 200μg/mL |
| Dibromomethane | 74-95-3 | 200μg/mL |
| Dichlorodifluoromethane | 75-71-8 | 200μg/mL |
| Ethylbenzene | 100-41-4 | 200μg/mL |
| Hexachlorobutadiene | 87-68-3 | 200μg/mL |
| Isopropylbenzene | 98-82-8 | 200μg/mL |
| Methylene chloride | 75-09-2 | 200μg/mL |
| m-Xylene | 108-38-3 | 200μg/mL |
| Naphthalene | 91-20-3 | 200μg/mL |
| n-Butylbenzene | 104-51-8 | 200μg/mL |
| n-Propylbenzene | 103-65-1 | 200μg/mL |
| o-Xylene | 95-47-6 | 200μg/mL |
| p-Isopropyltoluene | 99-87-6 | 200μg/mL |
| p-Xylene | 106-42-3 | 200μg/mL |
| sec-Butylbenzene | 135-98-8 | 200μg/mL |
| Styrene | 100-42-5 | 200μg/mL |
| tert-Butylbenzene | 98-06-6 | 200μg/mL |
| Tetrachloroethene | 127-18-4 | 200μg/mL |
| Toluene | 108-88-3 | 200μg/mL |
| trans-1,2-Dichloroethene | 156-60-5 | 200μg/mL |
| trans-1,3-Dichloropropene | 10061-02-6 | 200μg/mL |
| Trichloroethene | 79-01-6 | 200μg/mL |
| Trichlorofluoromethane | 75-69-4 | 200μg/mL |
| Vinyl Chloride | 75-01-4 | 200μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Specifications
| Chemical Name or Material | Volatile Organics Mix (Low Level) |
| Packaging | 2mL amber prescored glass ampule |
| Concentration or Composition Notes | Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity. |
| CAS Max % | 98.8, 0.02, 0.02, 0.02, 0.02, 0.02 |
| CAS | 87-68-3, 98-82-8, 108-38-3, 87-61-6, 96-18-4 |
| Methods | For US EPA Methods 502.2, 524, 624, 8260, 8260B |
| Analyte or Component Names | 1,1,1 |
| For Use With (Equipment) | GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation |
| Type | Certified Reference Material |
| Solvent or Matrix | Methanol-P and T |
| Show More |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product intended for laboratory use only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction