Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Volatiles Organics Combination Standard, SPEX CertiPrep™
Shop All Spex CertiPrep Products

Click to view available options
Quantity:
1 mL
Description
Includes
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Analyte | CAS No. | Concentration |
---|---|---|
1,1,1-Trichloroethane | 71-55-6 | 2000μg/mL |
1,1,2,2-Tetrachloroethane | 79-34-5 | 2000μg/mL |
1,1,2-Trichloroethane | 79-00-5 | 2000μg/mL |
1,1-Dichloroethane | 75-34-3 | 2000μg/mL |
1,1-Dichloroethene | 75-35-4 | 2000μg/mL |
1,2-Dichloroethane | 107-06-2 | 2000μg/mL |
1,2-Dichloropropane | 78-87-5 | 2000μg/mL |
2-Butanone | 78-93-3 | 2000μg/mL |
2-Hexanone | 591-78-6 | 2000μg/mL |
4-Methyl-2-pentanone | 108-10-1 | 2000μg/mL |
Acetone | 67-64-1 | 2000μg/mL |
Benzene | 71-43-2 | 2000μg/mL |
Bromodichloromethane | 75-27-4 | 2000μg/mL |
Bromoform | 75-25-2 | 2000μg/mL |
Carbon Disulfide | 75-15-0 | 2000μg/mL |
Carbon tetrachloride | 56-23-5 | 2000μg/mL |
Chlorobenzene | 108-90-7 | 2000μg/mL |
Chloroform | 67-66-3 | 2000μg/mL |
cis-1,2-Dichloroethene | 156-59-2 | 2000μg/mL |
cis-1,3-Dichloropropene | 10061-01-5 | 2000μg/mL |
Dibromochloromethane | 124-48-1 | 2000μg/mL |
Ethylbenzene | 100-41-4 | 2000μg/mL |
Methylene chloride | 75-09-2 | 2000μg/mL |
m-Xylene | 108-38-3 | 2000μg/mL |
o-Xylene | 95-47-6 | 2000μg/mL |
p-Xylene | 106-42-3 | 2000μg/mL |
Styrene | 100-42-5 | 2000μg/mL |
Tetrachloroethene | 127-18-4 | 2000μg/mL |
Toluene | 108-88-3 | 2000μg/mL |
trans-1,2-Dichloroethene | 156-60-5 | 2000μg/mL |
trans-1,3-Dichloropropene | 10061-02-6 | 2000μg/mL |
Trichloroethene | 79-01-6 | 2000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Specifications
Chemical Name or Material | Volatile Organics Combination Standard |
Packaging | 2mL amber prescored glass ampule |
Concentration or Composition Notes | Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. |
Methods | For US EPA CLP Methods |
For Use With (Equipment) | GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation |
Solvent or Matrix | Methanol-P and T |
Quantity | 1 mL |
Product intended for laboratory use only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction