Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Von Hippel Lindau Antibody (OTI1E1) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274851
Description
Von Hippel Lindau Monoclonal specifically detects Von Hippel Lindau in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Von Hippel Lindau | |
Monoclonal | |
LYOPH | |
Western Blot 1:4000, Immunohistochemistry 1:150 | |
elongin binding protein, HRCA1, Protein G7, pVHL, RCA1, VHL1, von Hippel-Lindau disease tumor suppressor, von Hippel-Lindau syndrome, von Hippel-Lindau tumor suppressor | |
Mouse | |
24 kDa | |
100 μg | |
Primary | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG2b |
Western Blot, Immunohistochemistry | |
OTI1E1 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
VHL | |
Full length human recombinant protein of human VHL(NP_000542) produced in HEK293T cell. | |
Affinity Purified | |
Apoptosis, Cancer, HIF Target Genes, Hypoxia, Neuronal Cell Markers, Neuronal Stem Cell Markers | |
7428 | |
Human, Mouse, Rat | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction