Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Von Willebrand Factor Antibody (F8/44/20), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250362F
Description
Von Willebrand Factor Monoclonal specifically detects Von Willebrand Factor in Human samples. It is validated for ELISA, Immunohistochemistry.Specifications
vWF-A2 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
VWF | |
Von Willebrand factor isolated from human plasma | |
0.1 mL | |
Primary | |
Human | |
Purified |
ELISA, Immunohistochemistry | |
F8/44/20 | |
ELISA, Immunohistochemistry | |
F8VWF, VWD, VWF von Willebrand factor | |
Mouse | |
Protein A purified | |
RUO | |
7450 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction