Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Von Willebrand Factor Antibody (F8/86) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. p-7079166 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
200 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB254379100 100 μg
NB254379200 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB254379100 Supplier Novus Biologicals Supplier No. NBP254379100UG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Von Willebrand Factor Monoclonal specifically detects Von Willebrand Factor in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.

Specifications

Antigen vWF-A2
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF
Classification Monoclonal
Clone F8/86
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml, CyTOF-ready
Formulation PBS with No Preservative
Gene Alias F8VWF, VWD, VWF von Willebrand factor
Gene Symbols VWF
Host Species Mouse
Immunogen von Willebrand Factor isolated from human plasma
Molecular Weight of Antigen 250 kDa
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7450
Target Species Human
Content And Storage Store at -20 to -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.